Gene Transcripts

Gene Symbol
SH2D7
Entrez Gene ID
646892
Gene Name
SH2 domain containing 7
Gene Aliases
Chromosome Location
Chr. 15: 78089895 - 78104051 on Build GRCh38
UniGene ID
Hs.569511
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101404.1 NP_001094874.1 5 1289
XM_011521895.2 XP_011520197.1 5 2581
XM_011521896.2 XP_011520198.1 6 1265
XM_011521899.2 XP_011520201.1 4 2056
XM_017022487.1 XP_016877976.1 6 1498
XM_017022488.1 XP_016877977.1 4 1073

View Full Product Details