Gene Transcripts

Gene Symbol
TPTE
Entrez Gene ID
7179
Gene Name
transmembrane phosphatase with tensin homology
Gene Aliases
CT44, PTEN2
Chromosome Location
Chr. 21: 10521514 - 10606270 on Build GRCh38
UniGene ID
Hs.122986
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290224.1 NP_001277153.1 19 1668
NM_199259.3 NP_954868.1 23 1890
NM_199260.3 NP_954869.1 22 1830
NM_199261.3 NP_954870.2 24 1944
GenBank mRNA AF007118.1 - 24 1892
AF495908.1 - 23 1838
AK308254.1 - 23 1787
AK314285.1 - 24 1843
AL833345.1 - 24 1851
AY219886.1 - 23 1838
AY219887.1 - 22 1778
AY219888.1 - 19 1616
BC028719.1 - 23 1873 5269598

View Full Product Details