Gene Transcripts

Gene Symbol
ZBTB2
Entrez Gene ID
57621
Gene Name
zinc finger and BTB domain containing 2
Gene Aliases
ZNF437
Chromosome Location
Chr. 6: 151364115 - 151391705 on Build GRCh38
UniGene ID
Hs.520073
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020861.2 NP_065912.1 1 100
XM_017011137.1 XP_016866626.1 1 105
GenBank mRNA AK291942.1 - 1 43
DB215154.1 - 1 101

View Full Product Details