Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000828.4 | NP_000819.3 | 11 | 1975 | |
NM_007325.4 | NP_015564.4 | 11 | 1975 | ||
GenBank mRNA | AK297136.1 | - | 11 | 1661 | |
BC117464.1 | - | 11 | 1847 | 40126015 | |
U10301.1 | - | 11 | 1735 | ||
U10302.1 | - | 11 | 1735 |