Gene Transcripts

Gene Symbol
Anxa7
Entrez Gene ID
11750
Gene Name
annexin A7
Gene Aliases
AI265384, AI316497, Anx7, synexin
Chromosome Location
Chr. 14: 20455261 - 20480133 on Build GRCm38
UniGene ID
Mm.280231
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110794.1 NP_001104264.1 10, 11 1331
NM_009674.3 NP_033804.2 10, 11 1446
GenBank mRNA AK032013.1 - 10, 11 1222
AK077579.1 - 10, 11 1139
AK134589.1 - 11, 12 1216
AK149654.1 - 10, 11 1138
AK151438.1 - 10, 11 1140
AK161613.1 - 10, 11 1140
AK167588.1 - 10, 11 1114
AK169199.1 - 10, 11 1113
AK217456.1 - 1, 2 111
BC008997.1 - 10, 11 1130 3157926
L13129.1 - 10, 11 1133

View Full Product Details