Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271353.1 | NP_001258282.1 | 9 | 1125 | |
NM_144855.3 | NP_659104.1 | 9 | 1128 | ||
NM_178224.3 | NP_835742.1 | 9 | 1128 | ||
GenBank mRNA | AK077669.1 | - | 9 | 1089 | |
AK085692.1 | - | 9 | 1092 | ||
BC013472.1 | - | 9 | 1054 | 4221477 | |
BC013480.1 | - | 9 | 1064 | 4239757 | |
BC022144.1 | - | 9 | 1089 | ||
BC026595.1 | - | 9 | 1061 | 4974777 |