Gene Transcripts

Gene Symbol
Col4a4
Entrez Gene ID
12829
Gene Name
collagen, type IV, alpha 4
Gene Aliases
E130010M05Rik, [a]4(IV)
Chromosome Location
Chr. 1: 82448419 - 82586849 on Build GRCm38
UniGene ID
Mm.40253
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007735.2 NP_031761.1 23 2012
GenBank mRNA AF169388.1 - 23 1955
AK164732.1 - 23 2013

View Full Product Details