Gene Transcripts
- Gene Symbol
- Cox5b
- Entrez Gene ID
- 12859
- Gene Name
- cytochrome c oxidase subunit Vb
- Gene Aliases
- Chromosome Location
- Chr. 1: 36691487 - 36693388 on Build GRCm38
- UniGene ID
- Mm.180182
- Species
- Mus musculus
- Gene Symbol
- Gm11273
- Entrez Gene ID
- 100040340
- Gene Name
- cytochrome c oxidase, subunit Vb pseudogene
- Gene Aliases
- OTTMUSG00000000426
- Chromosome Location
- Chr. 13: 21500935 - 21501452 on Build GRCm38
- UniGene ID
- Mm.485752
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC023228.1 |
- |
|
282 |
|
View Full Product Details