Gene Transcripts

Gene Symbol
Cys1
Entrez Gene ID
12879
Gene Name
cystin 1
Gene Aliases
2900006B19Rik, AV218859, Ccap, ck, cpk
Chromosome Location
Chr. 12: 24665833 - 24681795 on Build GRCm38
UniGene ID
Mm.52265
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004455.2 NP_001004455.2 5 1514
NM_001161807.1 NP_001155279.1 5 1554
NM_138686.3 NP_619627.3 5 1585
GenBank mRNA AF390548.1 - 5 1603
AK013490.1 - 5 746
BC139298.1 - 5 782 8862320
BC139302.1 - 5 742 8862324
BC145598.1 - 5 782 9054164
BC171971.1 - 5 742 9054257
BE996554.1 - 5 488
CF613278.1 - 1 233

View Full Product Details