Gene Transcripts

Gene Symbol
Klk1b9
Entrez Gene ID
13648
Gene Name
kallikrein 1-related peptidase b9
Gene Aliases
Egfbp-3, Egfbp3, Klk9, mGk-9
Chromosome Location
Chr. 7: 43976061 - 43980376 on Build GRCm38
UniGene ID
Mm.440884
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010116.1 NP_034246.1 525
GenBank mRNA BC024624.1 - 504
BC048869.1 - 489 4164261
M17962.1 - 525

View Full Product Details