Gene Transcripts

Gene Symbol
Cers1
Entrez Gene ID
93898
Gene Name
ceramide synthase 1
Gene Aliases
Lass1, Uog-1, to
Chromosome Location
Chr. 8: 70315775 - 70331588 on Build GRCm38
UniGene ID
Mm.258280
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138647.3 NP_619588.1 8 2660
GenBank mRNA AK053885.1 - 8 2665

Gene Symbol
Gdf1
Entrez Gene ID
14559
Gene Name
growth differentiation factor 1
Gene Aliases
AI385651, Gdf-1
Chromosome Location
Chr. 8: 70315775 - 70331592 on Build GRCm38
UniGene ID
Mm.258280
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163282.2 NP_001156754.1 8 2660
NM_008107.4 NP_032133.2 2 1338
GenBank mRNA AK162823.1 - 8 2643
BB183489.1 - 1 112
BC079555.1 - 3 1589 30359238
BC094351.1 - 8 2612
M57639.1 - 2 1338
M62301.1 - 8 2648

View Full Product Details