Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010324.2 | NP_034454.2 | 9 | 1273 | |
GenBank mRNA | AK146445.1 | - | 9 | 1175 | |
AK147807.1 | - | 9 | 1196 | ||
AK147934.1 | - | 9 | 1177 | ||
BC002057.1 | - | 9 | 1161 | 3489628 | |
J02623.1 | - | 9 | 1177 |