Gene Transcripts

Gene Symbol
Ifit2
Entrez Gene ID
15958
Gene Name
interferon-induced protein with tetratricopeptide repeats 2
Gene Aliases
AV302338, Ifi54, P54
Chromosome Location
Chr. 19: 34550549 - 34576534 on Build GRCm38
UniGene ID
Mm.2036
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008332.3 NP_032358.1 3 1138
GenBank mRNA AK038027.1 - 3 891
AK042366.1 - 3 920
AK150206.1 - 2 740
AK150253.1 - 2 740
BC050835.1 - 3 886 6513417
CT010395.1 - 1 662
U43085.1 - 2 713

View Full Product Details