Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008496.4 | NP_032522.2 | 3 | 239 | |
GenBank mRNA | AF038562.1 | - | 164 | ||
AF331640.1 | - | 2 | 144 | ||
AK003153.1 | - | 3 | 173 | ||
AK008773.1 | - | 3 | 173 | ||
AK009151.1 | - | 3 | 182 | ||
BC116838.1 | - | 3 | 200 | 40126157 | |
BC116840.1 | - | 3 | 200 | 40126159 | |
BG803457.1 | - | 3 | 396 |