Gene Transcripts
- Gene Symbol
- Mmp12
- Entrez Gene ID
- 17381
- Gene Name
- matrix metallopeptidase 12
- Gene Aliases
- AV378681, Mmel
- Chromosome Location
- Chr. 9: 7344397 - 7360461 on Build GRCm38
- UniGene ID
- Mm.2055
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008605.3 |
NP_032631.3
|
1, 2 |
171 |
|
GenBank mRNA |
AK089295.1 |
- |
|
140 |
|
AK089309.1 |
- |
1, 2 |
140 |
|
AK089335.1 |
- |
1, 2 |
136 |
|
AK089452.1 |
- |
1, 2 |
140 |
|
AK089523.1 |
- |
1, 2 |
140 |
|
AK090051.1 |
- |
1, 2 |
142 |
|
AK150596.1 |
- |
1, 2 |
140 |
|
AK151021.1 |
- |
1, 2 |
140 |
|
AK151273.1 |
- |
1, 2 |
140 |
|
AK151933.1 |
- |
1, 2 |
140 |
|
AK155139.1 |
- |
1, 2 |
139 |
|
AK155743.1 |
- |
1, 2 |
140 |
|
AK155747.1 |
- |
1, 2 |
140 |
|
AK155748.1 |
- |
1, 2 |
140 |
|
AK155865.1 |
- |
1, 2 |
139 |
|
AK170360.1 |
- |
1, 2 |
140 |
|
AK170364.1 |
- |
1, 2 |
140 |
|
AK170798.1 |
- |
1, 2 |
139 |
|
AK170804.1 |
- |
1, 2 |
139 |
|
AK170976.1 |
- |
1, 2 |
140 |
|
AK171006.1 |
- |
1, 2 |
140 |
|
AK171032.1 |
- |
1, 2 |
139 |
|
AK171036.1 |
- |
1, 2 |
140 |
|
AK171176.1 |
- |
1, 2 |
140 |
|
AK171428.1 |
- |
1, 2 |
140 |
|
AK171447.1 |
- |
1, 2 |
140 |
|
AK171448.1 |
- |
1, 2 |
140 |
|
AK171449.1 |
- |
1, 2 |
140 |
|
AK171450.1 |
- |
1, 2 |
140 |
|
AK171466.1 |
- |
1, 2 |
140 |
|
AK171468.1 |
- |
1, 2 |
140 |
|
AK171470.1 |
- |
1, 2 |
140 |
|
AK171472.1 |
- |
1, 2 |
140 |
|
AK171485.1 |
- |
1, 2 |
140 |
|
AK171518.1 |
- |
1, 2 |
140 |
|
AK171542.1 |
- |
1, 2 |
140 |
|
AK171570.1 |
- |
1, 2 |
140 |
|
BC019135.1 |
- |
1, 2 |
134 |
5037674 |
CT010377.1 |
- |
1, 2 |
87 |
|
M82831.1 |
- |
1, 2 |
87 |
|
View Full Product Details