Gene Transcripts

Gene Symbol
Mmp15
Entrez Gene ID
17388
Gene Name
matrix metallopeptidase 15
Gene Aliases
AI503551, MT2-MMP
Chromosome Location
Chr. 8: 95352337 - 95374293 on Build GRCm38
UniGene ID
Mm.217116
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008609.3 NP_032635.1 7 1721
GenBank mRNA AY622975.1 - 2 128
BC047278.1 - 7 1721 6390113
BC057952.1 - 7 1291 5342023
D86332.1 - 7 1217

View Full Product Details