Gene Transcripts

Gene Symbol
Mtl5
Entrez Gene ID
17771
Gene Name
metallothionein-like 5, testis-specific (tesmin)
Gene Aliases
tesmin
Chromosome Location
Chr. 19: 3388857 - 3407823 on Build GRCm38
UniGene ID
Mm.209941
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039657.2 NP_001034746.1 4 813
NM_001039658.2 NP_001034747.1 4 813
NM_001286557.1 NP_001273486.1 5 897
NM_001286558.1 NP_001273487.1 4 828
GenBank mRNA AB057423.1 - 4 801
AF329359.1 - 4 838
AF329360.1 - 5 907
AK015732.1 - 4 786
AK161511.1 - 4 814
BC024377.1 - 4 789 4976492
U67176.1 - 6 830
U77383.1 - 4 450

View Full Product Details