Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008542.2 | NP_001008542.2 | 5 | 806 | |
NM_001008543.2 | NP_001008543.1 | 5 | 493 | ||
NM_010847.3 | NP_034977.1 | 5 | 646 | ||
GenBank mRNA | AK140465.1 | - | 5 | 596 | |
AK144219.1 | - | 5 | 688 | ||
AK154538.1 | - | 5 | 796 | ||
BC064453.1 | - | 5 | 791 | 6810025 | |
D31824.1 | - | 5 | 646 | ||
L38821.1 | - | 5 | 499 | ||
L38822.1 | - | 5 | 518 | ||
U24673.1 | - | 5 | 479 | ||
U24674.1 | - | 5 | 428 |