Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010879.3 | NP_035009.3 | 4 | 874 | |
GenBank mRNA | AF043260.1 | - | 3 | 511 | |
AK014949.1 | - | 4 | 867 | ||
AK051665.1 | - | 4 | 874 | ||
BC011071.1 | - | 2 | 439 | 3673483 | |
BC034255.1 | - | 4 | 614 | 3157491 |