Gene Transcripts

Gene Symbol
Nnat
Entrez Gene ID
18111
Gene Name
neuronatin
Gene Aliases
5730414I02Rik, AW107673, Peg5
Chromosome Location
Chr. 2: 157560078 - 157562525 on Build GRCm38
UniGene ID
Mm.34330
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291128.1 NP_001278057.1 3 1002
NM_001291129.1 NP_001278058.1 3 990
NM_001291130.1 NP_001278059.1 3 941
NM_010923.3 NP_035053.1 3 995
NM_180960.3 NP_851291.1 2 914
GenBank mRNA AB004048.1 - 3 1202
AK003004.1 - 3 959
AK160626.1 - 3 963
BC036984.1 - 3 953 4981517
BU705167.1 - 1 256
CJ302750.1 - 1 169
GQ184462.1 - 3 897
X83568.1 - 3 763
X83569.1 - 2 870
X83570.1 - 3 951

View Full Product Details