Gene Transcripts

Gene Symbol
IFNAR2
Entrez Gene ID
3455
Gene Name
interferon alpha and beta receptor subunit 2
Gene Aliases
IFN-R, IFN-alpha-REC, IFNABR, IFNARB, IMD45
Chromosome Location
Chr. 21: 33229895 - 33264513 on Build GRCh38
UniGene ID
Hs.708195
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000874.4 NP_000865.2 7 994
NM_001289125.1 NP_001276054.1 7 994
NM_001289126.1 NP_001276055.1 7 948
NM_001289128.1 NP_001276057.1 7 994
NM_207584.2 NP_997467.1 7 948
NM_207585.2 NP_997468.1 7 948
GenBank mRNA AK293059.1 - 6 631
BC002793.2 - 7 942 3626374
CR541817.1 - 6 589
HG380509.1 - 7 714
L41942.1 - 7 915
L41943.1 - 6 628
L41944.1 - 7 915
U29584.1 - 7 655
X77722.1 - 7 814
X89772.1 - 6 589
X89814.1 - 7 753

View Full Product Details