Gene Transcripts

Gene Symbol
Psma2
Entrez Gene ID
19166
Gene Name
proteasome (prosome, macropain) subunit, alpha type 2
Gene Aliases
Lmpc3
Chromosome Location
Chr. 13: 14613242 - 14625673 on Build GRCm38
UniGene ID
Mm.252255
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008944.2 NP_032970.2 3 169
GenBank mRNA AF099733.1 - 3 136
AK012119.1 - 3 151
AK012990.1 - 3 158
AK035578.1 - 158
AK136115.1 - 3 171
AK151883.1 - 3 160
AK152862.1 - 3 160
AK154011.1 - 3 153
AK167752.1 - 3 160
BC026768.1 - 3 142 3156907
CJ052982.1 - 3 171
X70303.1 - 3 213

View Full Product Details