Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011193.2 | NP_035323.2 | 9 | 1016 | |
GenBank mRNA | AK037936.1 | - | 8 | 1818 | |
AK088150.1 | - | 8 | 1818 | ||
BC096761.1 | - | 9 | 796 | 30040955 | |
U87814.1 | - | 9 | 856 |