Gene Transcripts

Gene Symbol
Tmsb4x
Entrez Gene ID
19241
Gene Name
thymosin, beta 4, X chromosome
Gene Aliases
Ptmb4, Tb4
Chromosome Location
Chr. X: 167207093 - 167213479 on Build GRCm38
UniGene ID
Mm.142729
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021278.2 NP_067253.1 3 573
GenBank mRNA AK010659.1 - 3 517
AK088783.1 - 3 517
AK162286.1 - 3 518
AK169108.1 - 3 520
AK169718.1 - 3 517
AK205579.1 - 1 298
BC018286.1 - 3 489 3501378
X16053.1 - 3 602

View Full Product Details