Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000588.3 | NP_000579.2 | 1 | 165 | |
GenBank mRNA | BC066272.1 | - | 1 | 158 | |
BC066273.1 | - | 1 | 158 | 6971774 | |
BC066274.1 | - | 1 | 158 | 6971775 | |
BC066275.1 | - | 1 | 158 | 6971776 | |
BC066276.1 | - | 1 | 158 | 6971777 | |
BC069472.1 | - | 1 | 161 | 7262122 | |
M14743.1 | - | 1 | 121 | ||
M17115.1 | - | 1 | 150 | ||
M20137.1 | - | 1 | 165 |