Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001173483.1 | NP_001166954.1 | 7 | 1017 | |
NM_020599.2 | NP_065624.1 | 7 | 1099 | ||
GenBank mRNA | AK014227.1 | - | 7 | 790 | |
AK140622.1 | - | 7 | 1018 | ||
BC016268.1 | - | 6 | 692 | 4481973 |