Gene Transcripts
- Gene Symbol
- Robo1
- Entrez Gene ID
- 19876
- Gene Name
- roundabout homolog 1 (Drosophila)
- Gene Aliases
- AW494633, AW742721, DUTT1, Gm310
- Chromosome Location
- Chr. 16: 72027553 - 73047491 on Build GRCm38
- UniGene ID
- Mm.310772
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019413.2 |
NP_062286.2
|
8 |
2092 |
|
GenBank mRNA |
Y17793.1 |
- |
8 |
1354 |
|
View Full Product Details