Gene Transcripts

Gene Symbol
Scnn1g
Entrez Gene ID
20278
Gene Name
sodium channel, nonvoltage-gated 1 gamma
Gene Aliases
[g]ENaC
Chromosome Location
Chr. 7: 121734420 - 121768477 on Build GRCm38
UniGene ID
Mm.35247
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011326.2 NP_035456.1 2 145
GenBank mRNA AF112187.1 - 2 145
BC021338.1 - 2 184 5042876

View Full Product Details