Gene Transcripts

Gene Symbol
Ccl12
Entrez Gene ID
20293
Gene Name
chemokine (C-C motif) ligand 12
Gene Aliases
MCP-5, Scya12
Chromosome Location
Chr. 11: 82101845 - 82103399 on Build GRCm38
UniGene ID
Mm.867
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011331.2 NP_035461.2 2 208
GenBank mRNA AF065934.1 - 2 174
AF065935.1 - 2 174
AF065936.1 - 2 174
AF065937.1 - 2 174
AF065938.1 - 2 174
AK012356.1 - 2 208
BC027520.1 - 2 163 1548072
U50712.1 - 2 207
U66670.1 - 2 184

View Full Product Details