Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019535.2 | NP_062408.2 | 4 | 323 | |
GenBank mRNA | AF326561.1 | - | 4 | 202 | |
AF326562.1 | - | 4 | 202 | ||
AK035210.1 | - | 654 | |||
AK043424.1 | - | 311 | |||
AK044370.1 | - | 4 | 338 | ||
AK047324.1 | - | 265 | |||
AK134970.1 | - | 2 | 34 | ||
BC018385.1 | - | 4 | 323 | 4503038 | |
U58886.1 | - | 4 | 286 |