Gene Transcripts

Gene Symbol
Snrnp70
Entrez Gene ID
20637
Gene Name
small nuclear ribonucleoprotein 70 (U1)
Gene Aliases
2700022N21Rik, 3200002N22Rik, AI325098, R74807, Rnu1p70, Rnulp70, Snrp70, Srnp70, U1-70, U1-70K
Chromosome Location
Chr. 7: 45376454 - 45395742 on Build GRCm38
UniGene ID
Mm.216386
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK005045.2 - 5 472
AK011564.1 - 5 428
AK012272.1 - 3 121
AK014311.2 - 5 469
AK077425.1 - 5 474
AK133115.1 - 5 473
AK146729.1 - 5 473
AK148013.1 - 5 499
BC049128.1 - 5 438
BC051414.1 - 4 273
BC131994.1 - 5 365 40130271
BC132020.1 - 5 365 40130297

View Full Product Details