Gene Transcripts

Gene Symbol
Vpreb1
Entrez Gene ID
22362
Gene Name
pre-B lymphocyte gene 1
Gene Aliases
CD179a, Vpreb-1
Chromosome Location
Chr. 16: 16868401 - 16869255 on Build GRCm38
UniGene ID
Mm.57574
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016982.2 NP_058678.1 2 667
GenBank mRNA AK010843.1 - 2 661
AK010879.1 - 2 661
BC117051.1 - 2 629 40126370
BC119175.1 - 1 716 8733924
BC144850.1 - 1 716 9053398
X05556.1 - 2 662

View Full Product Details