Gene Transcripts

Gene Symbol
Zfp26
Entrez Gene ID
22688
Gene Name
zinc finger protein 26
Gene Aliases
5033428C05Rik, KRAB15, Zfp-26, Zfp70, Zfp81-rs1, mKIAA4196, mkr-3, mszf14, mszf52
Chromosome Location
Chr. 9: 20428318 - 20460169 on Build GRCm38
UniGene ID
Mm.107441
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011753.3 NP_035883.2 10 2034
GenBank mRNA AK029298.1 - 8 2299
AK082002.1 - 10 1868
AK141740.1 - 9 2397
AK220539.1 - 11

View Full Product Details