Gene Transcripts

Gene Symbol
Zhx1
Entrez Gene ID
22770
Gene Name
zinc fingers and homeoboxes 1
Gene Aliases
Chromosome Location
Chr. 15: 58047003 - 58076508 on Build GRCm38
UniGene ID
Mm.244931
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042438.2 NP_001035903.1 4 1845
NM_009572.4 NP_033598.2 3 1719
GenBank mRNA AK051410.1 - 3 1708
AK053191.1 - 3 1429
AK215128.1 - 1 17
AK220508.1 - 3 1680
BC004723.1 - 4 1552
BC032067.1 - 1 31
BC054543.1 - 2 1303 6406286
Z54200.1 - 3 1430

View Full Product Details