Gene Transcripts

Gene Symbol
Magel2
Entrez Gene ID
27385
Gene Name
melanoma antigen, family L, 2
Gene Aliases
Mage-l2, NDNL1, nM15, ns7
Chromosome Location
Chr. 7: 62376979 - 62381640 on Build GRCm38
UniGene ID
Mm.45207
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013779.2 NP_038807.4 1 2980
GenBank mRNA AJ243608.1 - 1 495
AK082944.1 - 1 2953
BC054763.1 - 1 272
BC062913.1 - 3 2485
BC071222.1 - 1 758

View Full Product Details