Gene Transcripts

Gene Symbol
Snf8
Entrez Gene ID
27681
Gene Name
SNF8, ESCRT-II complex subunit, homolog (S. cerevisiae)
Gene Aliases
D11Moh34
Chromosome Location
Chr. 11: 96034890 - 96047430 on Build GRCm38
UniGene ID
Mm.372235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033568.2 NP_291046.1 6 633
GenBank mRNA AK013058.1 - 6 635
AK020203.1 - 4 323
AK180940.1 - 1 18
BC003938.1 - 6 605 3486703
BC030490.1 - 1 598

View Full Product Details