Gene Transcripts
- Gene Symbol
- Pttg1
- Entrez Gene ID
- 30939
- Gene Name
- pituitary tumor-transforming gene 1
- Gene Aliases
- AW555095, C87862, Pttg, Pttg3
- Chromosome Location
- Chr. 11: 43420248 - 43426248 on Build GRCm38
- UniGene ID
- Mm.6856
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001131054.1 |
NP_001124526.1
|
3, 4 |
327 |
|
NM_013917.2 |
NP_038945.2
|
2, 3 |
579 |
|
GenBank mRNA |
AF069051.1 |
- |
2, 3 |
571 |
|
AF071209.1 |
- |
2, 3 |
579 |
|
AK002473.1 |
- |
2, 3 |
293 |
|
AK008704.1 |
- |
2, 3 |
294 |
|
AK088387.1 |
- |
2, 3 |
293 |
|
BC023324.1 |
- |
3, 4 |
333 |
4946234 |
CN837307.1 |
- |
3, 4 |
332 |
|
DQ157177.1 |
- |
2, 3 |
268 |
|
DV657646.1 |
- |
2, 3 |
297 |
|
View Full Product Details