Gene Transcripts

Gene Symbol
Tfr2
Entrez Gene ID
50765
Gene Name
transferrin receptor 2
Gene Aliases
Trfr2
Chromosome Location
Chr. 5: 137569822 - 137588081 on Build GRCm38
UniGene ID
Mm.21757
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289507.1 NP_001276436.1 3 547
NM_001289509.1 NP_001276438.1 5 566
NM_001289511.1 NP_001276440.1 4 424
NM_015799.4 NP_056614.3 5 506
GenBank mRNA AF207741.1 - 3 381
AF222895.1 - 4 424
AK004848.2 - 5 482
AK004965.1 - 5 537
AK155526.1 - 4 644
AK165320.1 - 3 375
BC013654.1 - 3 547 4196597

View Full Product Details