Gene Transcripts

Gene Symbol
Atg13
Entrez Gene ID
51897
Gene Name
autophagy related 13
Gene Aliases
1110053A20Rik, D2Ertd391e, Harbi1
Chromosome Location
Chr. 2: 91674612 - 91710598 on Build GRCm38
UniGene ID
Mm.28492
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145528.3 NP_663503.1 5 655
GenBank mRNA AK122339.1 - 1 374
AK149672.1 - 5 614
AK154470.1 - 5 641
AK163962.1 - 5 607
AK171206.1 - 5 641
AK171440.1 - 5 632
BC016669.1 - 5 639 2648797
BC023673.1 - 5 605 5343236
BC023712.1 - 5 628 5346303
BC033419.1 - 5 606 5359944

View Full Product Details