Gene Transcripts

Gene Symbol
Atg13
Entrez Gene ID
51897
Gene Name
autophagy related 13
Gene Aliases
1110053A20Rik, D2Ertd391e, Harbi1
Chromosome Location
Chr. 2: 91674612 - 91710598 on Build GRCm38
UniGene ID
Mm.28492
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145528.3 NP_663503.1 3, 4 506
GenBank mRNA AK149672.1 - 3, 4 465
AK154470.1 - 3, 4 492
AK163962.1 - 3, 4 458
AK171206.1 - 3, 4 492
AK171440.1 - 3, 4 483
BC016669.1 - 3, 4 490 2648797
BC023673.1 - 3, 4 456 5343236
BC023712.1 - 3, 4 479 5346303
BC033419.1 - 3, 4 457 5359944

View Full Product Details