Gene Transcripts

Gene Symbol
Ankzf1
Entrez Gene ID
52231
Gene Name
ankyrin repeat and zinc finger domain containing 1
Gene Aliases
1300008P06Rik, 2810025E10Rik, D1Ertd161e
Chromosome Location
Chr. 1: 75192160 - 75199387 on Build GRCm38
UniGene ID
Mm.259326
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026187.5 NP_080463.3 9 1590
GenBank mRNA AK004948.2 - 7 2020
AK012672.1 - 2 163
AK156255.1 - 9 1552
AK157405.1 - 9 1552
BC039910.1 - 7 2100
BC057896.1 - 4 432
BC071240.1 - 9 1520
BC150879.1 - 9 1540 9087790

View Full Product Details