Gene Transcripts

Gene Symbol
Pmm2
Entrez Gene ID
54128
Gene Name
phosphomannomutase 2
Gene Aliases
AI585868, C86848
Chromosome Location
Chr. 16: 8637707 - 8657552 on Build GRCm38
UniGene ID
Mm.9699
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016881.2 NP_058577.1 2 172
GenBank mRNA AF043514.1 - 2 172
AK002858.1 - 2 144
AK003334.1 - 2 130
AK005074.1 - 2 154
AK012683.1 - 2 156
AK134850.1 - 124
AK145402.1 - 2 124
AK149116.1 - 2 134
AK162626.1 - 2 142
BC037101.1 - 2 149
BC046325.1 - 2 120 6395635
BC094448.1 - 2 138 4238664

View Full Product Details