Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023158.6 | NP_075647.3 | 4 | 991 | |
GenBank mRNA | AF277001.1 | - | 4 | 454 | |
AF301017.1 | - | 4 | 520 | ||
AK003085.1 | - | 4 | 650 | ||
AK009599.1 | - | 4 | 540 | ||
AK028875.1 | - | 4 | 993 | ||
AK030515.1 | - | 4 | 416 | ||
AK150224.1 | - | 4 | 679 | ||
AK151330.1 | - | 4 | 765 | ||
AK152102.1 | - | 4 | 765 | ||
AK152547.1 | - | 4 | 943 | ||
AK153192.1 | - | 4 | 707 | ||
AK170428.1 | - | 5 | 1162 | ||
BC019961.1 | - | 4 | 872 | 3988542 | |
GQ380016.1 | - | 5 | 557 | ||
GQ380017.1 | - | 6 | 608 |