Gene Transcripts

Gene Symbol
Mustn1
Entrez Gene ID
66175
Gene Name
musculoskeletal, embryonic nuclear protein 1
Gene Aliases
1110028G01Rik, Mustang
Chromosome Location
Chr. 14: 30879257 - 30881610 on Build GRCm38
UniGene ID
Mm.220895
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181390.3 NP_852055.1 3 552
GenBank mRNA AJ277212.1 - 3 688
AK003360.1 - 3 268
AK003945.1 - 3 270
BC116939.1 - 3 260 40126258
BC116941.1 - 3 260 40126260

View Full Product Details