Gene Transcripts

Gene Symbol
Rps21
Entrez Gene ID
66481
Gene Name
ribosomal protein S21
Gene Aliases
1810049N11Rik, 2410030A14Rik
Chromosome Location
Chr. 2: 180257373 - 180258444 on Build GRCm38
UniGene ID
Mm.289669
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025587.2 NP_079863.1 4, 5 272
GenBank mRNA AA049954.1 - 3, 4 198
AK007846.1 - 4, 5 235
AK007850.1 - 4, 5 227
AK010610.1 - 4, 5 273
AK010637.1 - 4, 5 240
AK012488.1 - 4, 5 235
BC027563.1 - 4, 5 220 3468174
BC086912.1 - 4, 5 237 6758965
BU534365.1 - 4, 5 275

View Full Product Details