Gene Transcripts

Gene Symbol
Ms4a4d
Entrez Gene ID
66607
Gene Name
membrane-spanning 4-domains, subfamily A, member 4D
Gene Aliases
2210417J23Rik
Chromosome Location
Chr. 19: 11536849 - 11558466 on Build GRCm38
UniGene ID
Mm.441056
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025658.4 NP_079934.2 6 786
GenBank mRNA AF237915.1 - 5 460
AK003110.1 - 6 828
AK008968.1 - 6 835
BC069842.1 - 6 786 30446709

View Full Product Details