Gene Transcripts

Gene Symbol
Nkapl
Entrez Gene ID
66707
Gene Name
NFKB activating protein-like
Gene Aliases
4921504I05Rik
Chromosome Location
Chr. 13: 21467047 - 21468501 on Build GRCm38
UniGene ID
Mm.116869
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025719.3 NP_079995.2 1 410
GenBank mRNA AK014812.1 - 1 397
AK015912.1 - 1 392
AK160424.1 - 1 212
BC048457.1 - 1 396
BC061080.1 - 1 402
BC089537.1 - 1 398
BC099550.1 - 1 410 30918538

View Full Product Details