Gene Transcripts

Gene Symbol
Tmx2
Entrez Gene ID
66958
Gene Name
thioredoxin-related transmembrane protein 2
Gene Aliases
2310042M24Rik, AA589631, Txndc14
Chromosome Location
Chr. 2: 84671311 - 84678828 on Build GRCm38
UniGene ID
Mm.371621
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290751.1 NP_001277680.1 5 593
NM_025868.4 NP_080144.1 7 893
GenBank mRNA AK009759.1 - 6 581
AK075750.1 - 7 895
AK145245.1 - 6 602
AK170587.1 - 6 590
BC013544.1 - 6 575 4242418
BC110994.1 - 6 576 5349078

View Full Product Details