Gene Transcripts

Gene Symbol
MAK
Entrez Gene ID
4117
Gene Name
male germ cell associated kinase
Gene Aliases
RP62
Chromosome Location
Chr. 6: 10762723 - 10838954 on Build GRCh38
UniGene ID
Hs.446125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242385.1 NP_001229314.1 7 916
NM_001242957.2 NP_001229886.1 6 842
NM_005906.5 NP_005897.1 6 842
NR_134935.1 7 895
NR_134936.1 7 984
XM_011514619.2 XP_011512921.1 7 981
XM_011514620.2 XP_011512922.1 7 1768
XM_011514621.2 XP_011512923.1 6 964
XM_011514622.2 XP_011512924.1 7 1293
XM_011514624.2 XP_011512926.1 1 35
XM_017010863.1 XP_016866352.1 7 1294
XM_017010864.1 XP_016866353.1 7 1292
XM_017010865.1 XP_016866354.1 6 964
XM_017010866.1 XP_016866355.1 7 947
GenBank mRNA AB593146.1 - 7 870
AF505623.1 - 6 842
AK097831.1 - 7 859
AK310329.1 - 7 859
AK314408.1 - 7 981
BC039825.1 - 6 849 5167621
JN226411.1 - 6 613

View Full Product Details