Gene Transcripts

Gene Symbol
MAP1B
Entrez Gene ID
4131
Gene Name
microtubule associated protein 1B
Gene Aliases
FUTSCH, MAP5, PPP1R102
Chromosome Location
Chr. 5: 72107268 - 72209570 on Build GRCh38
UniGene ID
Hs.335079
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324255.1 NP_001311184.1 4 5502
NM_005909.4 NP_005900.2 5 5772
GenBank mRNA CR933720.1 - 1 3420

View Full Product Details